Skip to Content
MilliporeSigma
All Photos(1)

Documents

MABT1579

Sigma-Aldrich

Anti-Cadherin 6 Antibody, clone 2B6-D8

Synonym(s):

Kidney cadherin;K-cadherin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2B6-D8, monoclonal

mol wt

calculated mol wt 88.31 kDa
observed mol wt ~120 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CDH6(1004)

General description

Cadherin-6 (UniProt: P55285; also known as Kidney cadherin, K-cadherin) is encoded by the CDH6 gene (Gene ID: 1004) in human. Cadherins are calcium-dependent cell adhesion proteins that preferentially interact with themselves in a homophilic manner in connecting cells. Cadherin-6 is a class II cadherin that is highly expressed in brain and kidney. Its lower expression is also reported in lung, pancreas, and gastric mucosa. It is involved in the morphogenesis of central nervous system and kidney. It is a single-pass type I membrane glycoprotein that is synthesized with a signal peptide (aa 1-18) and a propeptide (aa 19-53) that are subsequently cleaved off to generate the mature form that contains an extracellular domain (aa 54-615), a transmembrane domain (aa 616-636), and a cytoplasmic domain (aa 637-790). It has 5 cadherin domains. Three calcium ions are usually bound at the interface of each cadherin domain and rigidify the connections and thereby imparting a strong curvature to the full-length ectodomain. Cadherin-6 is reported to drive EMT during embryonic development. It acts as a metastasis suppressor molecule and its expression is significantly reduced with an increase in tumor grade and stage in renal cell carcinoma. (Ref.: Gugnoni, M., et al. (2017). Oncogene 36(5); 667-677; Shimazui, T., et al. (1998). Clin. Cancer Res. 4(10); 2419-2424).

Specificity

Clone 2B6-D8 is a mouse monoclonal antibody that detects human Cadherin-6. It targets an epitope within the extracellular domain.

Immunogen

GST-tagged recombinant protein fragment corresponding to the extracellular domain of human Cadherin-6.

Application

Quality Control Testing

Evaluated by Western Blotting in Human kidney tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Cadherin-6 in Human kidney tissue lysate.

Tested Applicaitions

Western Blotting Analysis: A representative lot detected Cadherin-6 in Western Blotting applications (Shimazui, T., et al. (1998). Clin Cancer Res. 4(10); 2419-24).

Immunohistochemistry Applications: A representative lot detected Cadherin-6 in Immunohistochemistry applications (Shimazui, T., et al. (1998). Clin Cancer Res. 4(10); 2419-24).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Cadherin 6, clone 2B6-D8, Cat. No. MABT1579, is a mouse monoclonal antibody that detects Cadherin-6 and is tested for use in Immunohistochemistry and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service