Skip to Content
MilliporeSigma

MABS1685

Anti-ARHGEF28 Antibody, clone 14C10.1

clone 14C10.1, from mouse

Synonym(s):

Rho guanine nucleotide exchange factor 28, 190 kDa guanine nucleotide exchange factor, p190-RhoGEF, p190RhoGEF, Rho guanine nucleotide exchange factor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.46
Conjugate:
unconjugated
Clone:
14C10.1, monoclonal
Application:
IHC, WB
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

14C10.1, monoclonal

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Rho guanine nucleotide exchange factor 28 (UniProt P0C6P5; also known as 190 kDa guanine nucleotide exchange factor, p190-RhoGEF, p190RhoGEF, Rho guanine nucleotide exchange factor) is encoded by the Arhgef28 (also known as Rgnef) gene (Gene ID 361882) in rat species. p190RhoGEF is an ubiquitously expressed protein that belongs to the same subclass of Dbl family GEFs as Lbc, Lfc, GEFH-1, and p115RhoGEF. It contains several potential regulatory motifs, including an N-terminal leucine-rich region, a cysteine-rich zinc-finger domain and a C-terminal region that may form an alpha-helical coiled-coil. p190RhoGEF binds to RhoA, but not with Rac and Cdc42. When overexpressed in neuronal cells, it induces cell rounding and inhibits neurite outgrowth. Endogenous p190RhoGEF is reported to localize to distinct RhoA-containing regions at the plasma membrane, to the cytosol and along microtubules. In vitro and in vivo binding experiments show that p190RhoGEF directly interacts with microtubules via its C-terminal region adjacent to the catalytic Dbl homology/pleckstrin homology domain. Ref.: Van Horck FP et al. (2001). J. Biol. Chem. 276, 4948-4956.
~190 kDa observed. 190.8 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

GST-tagged recombinant rat ARHGEF28 fragment encompassing the N-terminal portion of microtubles-interacting domain, excluding sequence C-terminal to the RNA-binding region.

Application

Detect ARHGEF28 using this mouse monoclonal Anti-ARHGEF28, clone 14C10.1, Cat. No. MABS1685, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected ARHGEF28 in rat heart and kidney tissue sections.

Biochem/physiol Actions

Clone 14C10.1 targets an epitope within the N-terminal portion of ARHGEF28 microtubles-interacting domain.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ARHGEF28 in 10 µg of PC12 rat pheochromocytoma cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library