Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA012878

Sigma-Aldrich

Anti-NLRP3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AGTAVPRL, Anti-AII, Anti-C1orf7, Anti-CIAS1, Anti-CLR1.1, FCAS, FCU, MWS, NALP3, PYPAF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

FKMHLEDYPPQKGCIPLPRGQTEKADHVDLATLMIDFNGEEKAWAMAVWIFAAINRRDLYEKAKRDEPKWGSDNARVSNPTVICQEDSIEEEWMGLLEYLSRISICKMKKDYRKKYR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NLRP3(114548)

General description

NLRP3 is a signaling protein belonging to leucine rich repeat family and regulates the apoptotic and inflammatory signaling pathways. Rabbit polyclonal anti- NLRP3 antibody can be used in western blotting. Anti- NLRP3 antibodies are specific for NLRP3 protein.

Immunogen

NACHT, LRR and PYD domains-containing protein 3 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76150

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeremy A Hirota et al.
American journal of respiratory cell and molecular biology, 49(4), 517-524 (2013-04-26)
Inflammasomes have emerged as playing key roles in inflammation and innate immunity. A growing body of evidence has suggested that the nucleotide-binding domain and leucine-rich repeat protein-3 (NLRP3) inflammasome is important in chronic airway diseases such as asthma and chronic
Lei Chen et al.
Cancer immunology, immunotherapy : CII (2023-01-01)
The NOD-like receptor family pyrin domain-containing 3 (NLRP3) inflammasome plays cell- and tissue-specific roles in cancer, meaning that its activation in different tumors or cells may play different roles in tumor progression. We have previously described the tumor-promoting function of
Ravichandran Panchanathan et al.
Oncotarget, 7(19), 28183-28194 (2016-04-09)
The molecular mechanisms by which hypoxia contributes to prostatic chronic inflammation (PCI) remain largely unknown. Because hypoxia stimulates the transcriptional activity of NF-κB, which "primes" cells for inflammasome activation by inducing the expression of NLRP3 or AIM2 receptor and pro-IL-1β
Heidi Hsieh et al.
Toxicology in vitro : an international journal published in association with BIBRA, 35, 24-30 (2016-05-18)
Zinc is both an essential and potentially toxic metal. It is widely believed that oral zinc supplementation can reduce the effects of the common cold; however, there is strong clinical evidence that intranasal (IN) zinc gluconate (ZG) gel treatment for
William O'Connor et al.
Journal of immunology (Baltimore, Md. : 1950), 171(12), 6329-6333 (2003-12-10)
Mutations in the cold-induced autoinflammatory syndrome 1 (CIAS1) gene have been recently linked to three chronic autoinflammatory disorders. These observations point to an important role for CIAS1 in regulating inflammatory processes. We report that TNF-alpha and ligands recognized by multiple

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service