Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22760

Sigma-Aldrich

Anti-STAT6 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22760

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STAT6(6778)

Immunogen

Immunogen Sequence: GI # 23397678, sequence 61-275
Recombinant signal transducer and activator of transcription 6

Application

Anti-STAT6 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

STAT6 is a human gene and is a member of the STAT family of transcription factors. It is mapped to human chromosome band 12q13. STAT6 has the same functional domain for both receptor coupling and dimerization. It is highly expressed in comparison with the other STAT proteins. STAT-6 silencing results in the reduction of HeLa cell programmed death and the expression of several important key factors related to programmed cell death. It is a highly sensitive and specific marker of solitary fibrous tumor (SFT). It can also be helpful in distinguishing tumor type from histologic mimics.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akihiko Yoshida et al.
The American journal of surgical pathology, 38(4), 552-559 (2014-03-15)
Solitary fibrous tumor (SFT) is an uncommon fibroblastic neoplasm. Although histologic characteristics and frequent CD34 expression allow for an accurate diagnosis in the majority of SFT cases, a wide histologic spectrum and an occasional unexpected immunophenotype may pose diagnostic challenges.
Zhibin Li et al.
Biomaterials, 35(18), 5016-5027 (2014-03-29)
Based on the fact that the transcription of STAT-1 plus its Serine 727 and Tyrosine 701 phosphorylation is not the pre-requisite for the cell death signal transduction in the IFN-γ signaling pathway induced by co-immobilized IFN-γ/TNF-α, we investigate both in
Leona A Doyle et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 27(3), 390-395 (2013-09-14)
Solitary fibrous tumor (SFT) is composed of spindled to ovoid cells in a patternless architecture with prominent stromal collagen and hemangiopericytoma-like vessels. Some tumors show hypercellularity, nuclear atypia, and significant mitotic activity; the latter feature in particular often portends an
Assignment of the STAT6 gene (STAT6) to human chromosome band 12q13 by in situ hybridization.
J P Leek et al.
Cytogenetics and cell genetics, 79(3-4), 208-209 (1997-01-01)
J Hou et al.
Science (New York, N.Y.), 265(5179), 1701-1706 (1994-09-16)
Interleukin-4 (IL-4) is an immunomodulatory cytokine secreted by activated T lymphocytes, basophils, and mast cells. It plays an important role in modulating the balance of T helper (Th) cell subsets, favoring expansion of the Th2 lineage relative to Th1. Imbalance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service