Skip to Content
MilliporeSigma
All Photos(1)

Documents

SCME004

Sigma-Aldrich

EndoGRO-MV Complete Culture Media Kit

Synonym(s):

EndoGRO Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100
NACRES:
NA.32

form

liquid

Quality Level

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

input

endothelial cells

General description

EndoGRO Media Formulations provide an optimal cell culture environment for many types of endothelial cells, including HUVEC, aortic endothelial cells and other human large vessel and microvascular endothelial cells. These media have been shown to grow endothelial cells at rates that meet or exceed commercially available serum containing media, while maintaining excellent cell morphology. The media are packaged in a specially designed UV protective shrinkwrap for added stability, which includes a temperature gauge that helps prevent contamination. EndoGRO Media contain no antimicrobials and no phenol red, components that can cause cell stress and masking effects that may influence experimental results. Four different media formulations are available for various applications and cell culture requirements.

Application

The EndoGRO-MV Culture Media kit is a 5% serum formulation approximately for culturing human microvascular endothelial cells. This media does not contain VEGF.

EndoGRO-MV is a 5% serum formulation approximately for culturing human microvascular endothelial cells. It does not contain VEGF.

Components

  • EndoGRO Basal Medium (SCME-BM)
  • Supplement Kit (SCME004-S)

Storage and Stability

Basal Medium (SCME-BM) should be stored at 2 - 8ºC. The special UV protective packaging protects EndoGRO Medium from light damage, however users should take care to protect medium from extended exposure to light. Supplement Kit (SCME004-S) should be stored at -20ºC.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service