Skip to Content
MilliporeSigma

GF305

IFN- γ Protein, Human Recombinant Animal Free

The Interferon gamma (or IFN-γ protein) is an acid-labile interferon produced by CD4 & CD8 T lymphocytes as well as activated NK cells. Manufactured using all non-animal reagents.

Synonym(s):

IFN, Interferon Gamma, IFNG, IFN Gamma

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
eCl@ss:
32160405
Assay:
>98% (SDS-PAGE and HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Interferon gamma (IFN-γ) is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-γ signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-γ stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Human IFN-γ species-specific and is biologically active only in human and primate cells. Recombinant human IFN-γ is a 16.7 kDa protein containing 143 amino acid residues.

Physical form

Recombinant animal free human IFN- γ is manufactured using all non-animal reagents

Analysis Note

Bioactivity assay: Determined by its ability to induce apoptosis in HeLa cells. The expected ED50 for this effect is 5.0-10.0 ng/ml.


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library