Skip to Content
MilliporeSigma
All Photos(1)

Documents

GE80-6483-94

Mini Dialysis Kit, 1 kDa

Cytiva 80-6483-94, pack of 2 mL

Synonym(s):

Mini Dialysis Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NA.25

feature

wetted part: no

packaging

pack of 2 mL

manufacturer/tradename

Cytiva 80-6483-94

storage temp.

2-8°C

General description

The disposable tubes in Mini Dialysis Kit offer a simple solution to the handling problems of small volume dialysis. The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane.

Application

Dialyzing small samples can be difficult and inefficient when the sample must be transferred into and out of dialysis bags and centrifuge tubes. The disposable tubes in Mini Dialysis Kit offer a simple solution to the handling problems of small volume dialysis.

The dialysis tubes in the kit each have a cap that incorporates a small disk of dialysis membrane. Samples are easily pipetted into and removed from the conical bottom of the tube. The capped tube is inserted into a float and then, with the cap and membrane down, placed in a stirred beaker containing the desired dialysis solution.

Following dialysis, the tube is centrifuged briefly. This brings the entire contents of the tube back to the conical bottom for maximum recovery. The dialyzed sample can be stored in the same tube by simply replacing the dialysis cap with a standard cap.

Features and Benefits

  • Designed for efficient dialysis of small sample volumes.
  • Uses disposable dialysis tubes with dialysis membrane incorporated into caps.
  • Conical tube bottom maximizes sample recovery

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.

Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.

Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.

Mini Dialysis Kit from Cytiva minimizes sample handling and loss, reducing streaking on 2-D gels from contaminants.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service