Documents
Z742661
Tube-O-Tank for use with the Tube-O-DIALYZER
size large
About This Item
UNSPSC Code:
41123100
NACRES:
NB.24
size
large
packaging
pkg of 1 ea
manufacturer/tradename
G-BioSciences 786-145E
General description
Dialysis tank specifically designed for use with the Tube-O-DIALYZER. Two sizes are available that are suitable for Micro (Small) and Medi (Large) size Tube-O-DIALYZER Each tank will accommodate 7 Tube-O-DIALYZER and are supplied with floats.
Legal Information
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.