Skip to Content
MilliporeSigma

Skip To

Z368024

Cellulose acetate membrane

dialyser capacity 500-1,500 μL, MWCO 500 Da, diam. 16 mm

Synonym(s):

Harvard Bioscience 7425-CA500, dialyzer filter, filter, membrane filter, particulate filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.22
UNSPSC Code:
41123100
Packaging:
pack of 25 ea
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 25 ea

dialyser capacity

500-1,500 μL

diam.

16 mm

pore size

500 Da MWCO

storage temp.

2-8°C

General description

Made of cellulose acetate for low protein binding. Nominal molecular weight limit (MWCO) is approximate upper limit of molecular weight to which membrane is permeable.

Application

Cellulose acetate membrane has been used in the removal of unreacted substances from the media. It has also been used in the ultrafiltration of water.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Z367982WHA10404106WHA10404139
description

dialyser capacity 500-1,500 μL, MWCO 500 Da, diam. 16 mm

description

dialyser capacity 10-200 μL, MWCO 500 Da, diam. 11 mm

description

cellulose acetate circles (OE 66), 0.2 μm, 25 mm 100/pk

description

OE66 membrane circles, cellulose acetate, pore size 0.2 μm, diam. 293 mm

packaging

pack of 25 ea

packaging

pack of 25 ea

packaging

pack of 100

packaging

pk of 25

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

dialyser capacity

500-1,500 μL

dialyser capacity

10-200 μL

dialyser capacity

-

dialyser capacity

-

pore size

500 Da MWCO

pore size

500 Da MWCO

pore size

0.2 μm

pore size

0.2 μm

diam.

16 mm

diam.

11 mm

diam.

25 mm

diam.

293 mm


Still not finding the right product?

Explore all of our products under Cellulose acetate membrane


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library