Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE80-1128-28

Immobiline® DryPlate precast gels kit

pH 4.0-7.0, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NC.01

feature

wetted part: no

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 80112828

pH

4.0-7.0

General description

Immobiline DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of NA.5 mm in the reswelling cassette accessory before use

Immobiline technology produces an absolutely stable pH gradient, allowing extremely high voltages and high resolution over narrow pH ranges, even for low mobility proteins that require long focusing times

Features and Benefits

  • Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients
  • Choose from narrow (1 pH unit) and medium (3 pH units) pH ranges
  • Handle mechanically stable, plastic-backed gels with confidence

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Immobiline is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service