Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE28-9440-09

NHS Mag Sepharose

Cytiva 28-9440-09, pack of 1 × 500 μL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

ligand

N-hydroxysuccinimide

packaging

pack of 1 × 500 μL

manufacturer/tradename

Cytiva 28-9440-09

storage condition

(2-propanol)

parameter

Room temperature temp. range

matrix

paramagnetic, spherical, highly cross-linked agarose particles

capacity

8- 14 μmol/mL binding capacity(gel)

storage temp.

2-8°C

General description

NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected proteins based on affinity.

Application

NHS Mag Sepharose are magnetic beads designed for pull-down techniques enabling rapid capture and enrichment of selected Proteins based on affinity. An antibody or other Protein with desired affinity is covalently coupled to the medium via primary amines for the purpose of capture of target Proteins of interest.

Features and Benefits

  • Visible and dense Sepharose based magnetic beads with Protein A ligand, easy to spot and collect bound target Protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target Protein in small or large sample volumes (Low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target Proteins - only Low amounts of antibody needed
  • Flexible protocols with elution conditions optimized for both electrophoresis and MS analysis

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Covalent coupling of antibodies, aptamers, and proteins with a NHS Mag Sepharose, Sera-Mag Carboxylate-Modified, Sera-Mag SpeedBeads Carboxylate-Modified from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service