Skip to Content
MilliporeSigma

Skip To

ROAHAHA

Roche

Anti-HA High Affinity

from rat IgG1

Synonym(s):

antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.54
Conjugate:
unconjugated
Clone:
3F10, monoclonal
Application:
Citations:
41
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3F10, monoclonal

form

lyophilized

packaging

pkg of 50 μg (11867423001), pkg of 500 μg (11867431001)

manufacturer/tradename

Roche

isotype

IgG1

epitope sequence

YPYDVPDYA

storage temp.

2-8°C

General description

Anti-HA High Affinity is a monoclonal antibody to the HA-peptide (clone 3F10). Anti-HA High Affinity recognizes the HA peptide sequence (YPYDVPDYA), derived from the influenza hemagglutinin protein. The antibody recognizes its antigenic determinant even when the HA peptide epitope is introduced into unrelated recombinant proteins by a technique known as "epitope tagging".

Immunogen

Amino acids 98-106 from the human influenza virus hemagglutinin protein

Application

Use Anti-HA High Affinity for the detection of native influenza hemagglutinin protein and recombinant proteins that contain the HA epitope using:
  • Dot blots
  • ELISA
  • Immunocytochemistry
  • Immunoprecipitation
  • Western blots
Since Anti-HA High Affinity is a rat monoclonal, it is possible to use it in conjunction with murine monoclonals for double labeling.

Biochem/physiol Actions

Anti-HA, High Affinity recognizes the 9-amino acid sequence YPYDVPDYA, derived from the human influenza hemagglutinin (HA) protein. This epitope is also recognized in fusion proteins regardless of its position (N-terminal, C-terminal or internal).

Preparation Note

Working concentration: Working concentration of conjugate depends on application and substrate.

The following concentrations should be taken as a guideline:
  • ELISA: for detection 100 ng/ml; for coating 1 to 5 μg/ml
  • Immunoprecipitation: 0.5 to 5 μg/ml
  • Western blot: 50 to 200 ng/ml

Add 0.5 ml (11 867 423 001; 50 μg) or 2.5 ml (11 867 431 001; 500 μg) double-distilled water to a final concentration of 100 μg/ml or 200 μg/ml respectively.
Rehydrate for 10 min prior to use.

Analysis Note

Function tested in western blot.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ROAHAH3663H6908
description

from rat IgG1

description

from mouse IgG2bκ

description

clone HA-7, purified from hybridoma cell culture

description

affinity isolated antibody, buffered aqueous solution

biological source

rat

biological source

mouse

biological source

mouse

biological source

rabbit

clone

3F10, monoclonal

clone

12CA5, monoclonal

clone

HA-7, monoclonal

clone

polyclonal

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

form

lyophilized

form

frozen liquid (11666606001), lyophilized (11583816001)

form

-

form

buffered aqueous solution

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


pictograms

Exclamation markExclamation mark

signalword

Warning-Warning

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library