Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-5316-02

Capto Q

Cytiva 17-5316-02, pack of 100 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pack of 100 mL

manufacturer/tradename

Cytiva 17-5316-02

matrix

highly cross-linked agarose with dextran surface extender

average diameter

90 μm (d50v)

cleaning in place

2-14

working range

2-12

suitability

suitable for bioprocess medium

General description

Capto Q is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective capture and intermediate protein purification.

Capto Q is designed for capture and intermediate purification of proteins from large feed volumes. It is composed of a rigid, high-flow agarose matrix modified with dextran surface extenders and a strong quaternary ammonium (Q) anion exchanger. The combination of high volume throughput and high dynamic binding capacity for Capto Q raises the productivity in industrial downstream processes. Capto Q withstands effective and rigorous CIP procedure and the chemical stability ensures long media lifetime.

Capto Q is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development.
As member of the BioProcess media range, Capto Q meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Strong anion exchanger for high-productivity capture and intermediate purification when high volume throughput is essential.
  • High dynamic binding capacity at high flow raises productivity
  • High-volume throughput cuts process times
  • Cost-effective processing with smaller unit operations.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Capto is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Description of viral clearance using Capto adhere from Cytiva.

Capto adhere from Cytiva is a multimodal strong anion exchanger for BioProcess applications.

This page describes the difference between bind/elute and flowthrough modes in multimodal chromatography using Cytiva products.

This page describes the maintenance of media and storage conditions for multimodal chromatography using Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service