Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GE17-1153-01

HiTrap® Q High Performance

Cytiva 17-1153-01, pack of 5 × 1 mL

Synonym(s):

Q-Sepharose High Performance

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-1153-01

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning

1-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

General description

HiTrap® Q HP are prepacked, ready-to-use Q Sepharose High Performance strong anion exchange columns for high-resolution, small-scale protein purification.

Application

Techniques & Applications: Separation of protein using a strong anion exchange medium prepacked in the HiTrap® column format.

Features and Benefits

  • 34 μm bead size for high-performance, high-resolution purifications.
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series.
  • Designed for use with a syringe, peristaltic pump, and chromatography systems such as AKTA design.
  • Strong quaternary ammonium (Q) anion exchanger

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to perform a purification of His-tagged membrane proteins.

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

Protocols

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service