Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

GE29-0513-24

HiTrap® SP HP

Cytiva 29-0513-24, pack of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

sulphopropyl

description

Ion Exchanger Type (value)

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 29-0513-24

availability

not available in North America

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

24-44 μm

avg. part. size

34 μm

cleaning

3-14

working range

4-13

suitability

suitable for bioprocess medium

General description

HiTrap SP HP are prepacked, ready-to-use SP Sepharose High Performance strong cation exchange columns for high-resolution, small-scale protein purification.

Application

The small particle size (34 μm) of the medium allows fast adsorption and desorption even at high sample loadings and flow rates.SP Sepharose High Performance is a strong cation exchange medium and Q Sepharose High Performance is a strong anion exchange medium. Both remain charged and have high loading capacities over broad pH ranges.

Features and Benefits

  • 34 μm bead size for high-performance, high-resolution purifications.
  • Convenient and reproducible for fast, easy, high-performance separations either alone or connected in series.
  • Designed for use with a syringe, peristaltic pump, and chromatography systems such as AKTA design.
  • Strong sulfopropyl (SP) cation exchanger.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to perform a purification of His-tagged membrane proteins.

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

Protocols

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

Ion exchange chromatography and Superdex column analysis facilitate protein refolding with step-by-step guidance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service