Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G9048

Sigma-Aldrich

GANT61

≥98% (HPLC)

Synonym(s):

2,2′-[[Dihydro-2-(4-pyridinyl)-1,3(2H,4H)-pyrimidinediyl]bis(methylene)]bis[N,N-dimethyl-benzenamine, NSC 136476

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H35N5
CAS Number:
Molecular Weight:
429.60
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >2 mg/mL

storage temp.

2-8°C

SMILES string

N2(C(N(CCC2)Cc4c(cccc4)N(C)C)c3ccncc3)Cc1c(cccc1)N(C)C

InChI

1S/C27H35N5/c1-29(2)25-12-7-5-10-23(25)20-31-18-9-19-32(27(31)22-14-16-28-17-15-22)21-24-11-6-8-13-26(24)30(3)4/h5-8,10-17,27H,9,18-21H2,1-4H3

InChI key

KVQOGDQTWWCZFX-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

GANT61 blocks hedgehog pathway signaling by inhibiting the transcripional activity of Gli1 (IC50 = 5 μM). GANT 61 led to a regression of 22Rv1 human prostate tumor cell xenografts in nude mice.
GANT61 is a Hh pathway Gli Inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syeda Kiran Riaz et al.
Scientific reports, 9(1), 6620-6620 (2019-05-01)
Sonic Hedgehog signaling is critical for breast morphogenesis and cancer. The present study was conducted to explore the influence of SHH/GLI1 axis on epithelial mesenchymal transition and invasion in breast cancer cells. SHH/GLI1 positive samples demonstrated high expression of Snail
Syeda Kiran Riaz et al.
Frontiers in cell and developmental biology, 9, 758400-758400 (2021-11-02)
The underlying mechanism of fibroblast growth factor receptor 1 (FGFR1) mediated carcinogenesis is still not fully understood. For instance, FGFR1 upregulation leads to endocrine therapy resistance in breast cancer patients. The current study aimed to identify FGFR1-linked genes to devise
Yonghong Hu et al.
Frontiers in pharmacology, 13, 1033103-1033103 (2022-12-10)
Backgroud and aims: Ductular reaction (DR) is a common pathological change and thought to have a key role in the pathogenesis and progression of liver fibrosis. Our previous study reported Gypenosides (GPs) ameliorated liver fibrosis, however, the anti-fibrotic mechanisms of
Taghi Manshouri et al.
Cell death & disease, 13(5), 481-481 (2022-05-21)
Bone marrow (BM) fibrosis was thought to be induced exclusively by mesenchymal stromal cells (MSCs). However, we and others found that neoplastic fibrocytes induce BM fibrosis in myelofibrosis (MF). Because glioma-associated oncogene-1 (GLI1), an effector of the Hedgehog pathway, plays
Chiachen Chen et al.
Cellular signalling, 46, 83-91 (2018-03-05)
Neuroendocrine (NE) lung tumors account for 20% of total lung cancer cases and represent a subset of aggressive tumors with metastatic potential. High-risk NE lung cancer patients display disseminated disease, N-myc expression/amplification, and poorly differentiated tumors. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service