Skip to Content
MilliporeSigma
All Photos(2)

Documents

344355

Sigma-Aldrich

AS1842856

≥98% (HPLC), solid, Foxo1 inhibitor, Calbiochem®

Synonym(s):

Foxo1 Inhibitor, AS1842856, 5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22FN3O3
CAS Number:
Molecular Weight:
347.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Foxo1 Inhibitor, AS1842856, Foxo1 Inhibitor, AS1842856, is a cell-permeable inhibitor that blocks the transcription activity of Foxo1 (IC₅₀ = 33 nM). Directly binds to the active Foxo1, but not the Ser256-phosphorylated form.

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white to tan
tan to yellow

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C1C(C(O)=O)=CN(C2=C1C(N)=C(F)C(NC3CCCCC3)=C2)CC

General description

A cell-permeable oxodihydroquinoline that preferentially inhibits the transcription activity of Forkhead box O family member Foxo1 (IC50 = 33 nM) over that of the functionally related Foxo3a and Foxo4 (70%, 20%, and 3% inhibition, respectively, in HepG2-based reporter assays; [AS184256] = 100 nM) via direct binding of the active Foxo1, but not the Ser256-phosphorylated/inactive form of Foxo1. Shown to inhibit gluconeogenesis in rat heptoma Fao cultures (IC50 = 43 nM against glucose production) in vitro and in liver of both non-diabetic and diabetic mice during a 26-hour fasting period (100 mg/kg/8 h p.o.) in vivo. Also available as a 50 mM solution in DMSO (Cat. No. 506081).
A cell-permeable oxodihydroquinoline that preferentially inhibits the transcription activity of Forkhead box O family member Foxo1 over that of the functionally related Foxo3a and Foxo4 (70%, 20%, and 3% inhibition, respectively, in HepG2-based reporter assays; [AS184256] = 100 nM) in a dose-dependent manner (IC50 = 33 nM) via direct binding of the non-Ser256-phosphorylated/active Foxo1, but not the Ser256-phosphorylated/inactive form of Foxo1. Shown to inhibit gluconeogenesis in rat heptoma Fao cultures (IC50 = 37 and 130 nM against PEPCK and G6Pase mRNA level; IC50 = 43 nM against glucose production) in vitro and in murine liver of both normoglycemic ICR mice (60% and 45% inhibition of hepatic G6Pase and PEPCK mRNA level, respectively; three 100 mg/kg p.o. dosages at 0, 12, 24 h) and diabetic db/db mice (86%, 70%, 56%, 48%, 35%, and 31% inhibition of hepatic G6Pase, ABCG8, ABCG5, PEPCK, IL-1β, and apoCIII mRNA level, respectively; three 100 mg/kg p.o. dosages at 0, 12, 24 h) during a 26-hour fasting period in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Nagashima, T., et al. 2010. Mol. Pharmacol.78, 961.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Dudek et al.
Cell reports, 38(7), 110389-110389 (2022-02-17)
Liver sinusoidal endothelial cells (LSECs) are liver-resident antigen (cross)-presenting cells that generate memory CD8 T cells, but metabolic properties of LSECs and LSEC-primed CD8 T cells remain understudied. Here, we report that high-level mitochondrial respiration and constitutive low-level glycolysis support LSEC scavenger
Matthew R Brown et al.
Science advances, 7(51), eabg6856-eabg6856 (2021-12-16)
Circadian rhythm disruption (CD) is associated with impaired glucose homeostasis and type 2 diabetes mellitus (T2DM). While the link between CD and T2DM remains unclear, there is accumulating evidence that disruption of fasting/feeding cycles mediates metabolic dysfunction. Here, we used
Xiangyu Zhang et al.
Circulation research, 133(3), 200-219 (2023-06-23)
The mTOR (mechanistic target of rapamycin) pathway is a complex signaling cascade that regulates cellular growth, proliferation, metabolism, and survival. Although activation of mTOR signaling has been linked to atherosclerosis, its direct role in lesion progression and in plaque macrophages
Xinxing Wang et al.
Aging, 13(8), 11969-11987 (2021-04-24)
Circular RNAs (circRNAs) have critical regulatory roles in tumor biology. However, their contributions in hepatocellular carcinoma (HCC) still remain enigmatic. The present study aimed to investigate the molecular mechanisms underlying the involvement of hsa_circ_0110102 in the occurrence and development of
Tomas Duffy et al.
Arthritis & rheumatology (Hoboken, N.J.), 72(9), 1514-1523 (2020-04-14)
Osteoarthritis (OA) is the most common age-related joint disease. With aging and in OA, the expression of FoxO transcription factors is reduced, diminishing their chondroprotective actions. In order to elucidate the molecular mechanisms by which FoxO1 protects chondrocytes, we sought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service