Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

G4423

Sigma-Aldrich

β-Glucanase from Trichoderma longibrachiatum

greener alternative

Synonym(s):

endo-1,3(4)-β-glucanase, endoglucanase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

Quality Level

specific activity

≥1.0 units/mg solid

composition

protein, ≥10%

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

General description

β -Glucanase represents a group of carbohydrate enzymes which break down glycosidic bonds within beta-glucan. It forms the main constituent of fungal cell walls and could be a potential structural and storage polysaccharide of marine macro-algae. It has the ability to degrade fungal cell walls and may be involved in defense mechanism of plants against pathogenic fungi.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles.

Application

β -Glucanase was used as a cellulase enzyme in the combined biological and chemical pretreatment method for lignocellulosic ethanol production from energy cane. It was also used in the enzymatic saccharification of cellulose and production of ethanol.

Biochem/physiol Actions

A mixture composed mainly of β-1→3 / 1→4-glucanase, xylanase, and cellulase activities. β-glucosidase, β-xylosidase, α-L-arabinofuranosidase, amylase, and protease activities are also present.

Unit Definition

One unit will liberate 1.0 μmole of glucose from cellulose in one hr at pH 5.0 at 37 °C (2 hr incubation time).

Physical form

contains maltodextrin, silica and sodium benzoate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Adams
Microbiology (Reading, England), 150(Pt 7), 2029-2035 (2004-07-17)
The fungal cell wall is a complex structure composed of chitin, glucans and other polymers, and there is evidence of extensive cross-linking between these components. The wall structure is highly dynamic, changing constantly during cell division, growth and morphogenesis. Hydrolytic
Combined biological and chemical pretreatment method for lignocellulosic ethanol production from energy cane;
Sara Shields, Raj Boopathy
Renewable Bioresources null
Ravindra Bansal et al.
Current genetics, 65(4), 953-964 (2019-03-07)
Trichoderma spp. are widely used as commercial biofungicides, and most commercial formulations are conidia based. Identification of genes that regulate conidiation would thus be of help in genetic reprogramming of these species to optimize sporulation. In this study, we constructed
Marie-Therese Eckstein et al.
Current biology : CB, 30(23), 4799-4807 (2020-10-10)
The human gut microbiota is composed of diverse microbes that not only compete but also rely on one another for resources and access to microhabitats in the intestine [1, 2]. Indeed, recent efforts to map the microbial biogeography of the
Juntao Sun et al.
Biotechnology letters, 33(11), 2193-2199 (2011-07-08)
A new fusion gene (Bgl-licMB), encoding β-1,3-1,4-glucanase both from Bacillus amyloliquefaciens (Bgl) and Clostridium thermocellum (licMB), was constructed via end-to-end fusion and expressed in Escherichia coli to improve hydrolytic activity and thermostability of β-1,3-1,4-glucanase. The results of enzymatic properties showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service