Skip to Content
MilliporeSigma

E2164

endo-1,4-β-D-glucanase from Acidothermus cellulolyticus

greener alternative

recombinant, expressed in corn, ≥2.0 units/mg protein

Synonym(s):

Endoglucanase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
Specific activity:
≥2.0 units/mg protein
Recombinant:
expressed in corn
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in corn

Quality Segment

specific activity

≥2.0 units/mg protein

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Physical form

Supplied as a suspension in 50 mM sodium acetate pH 5 in 3.2 M ammonium sulfate; 0.02% sodium azide

Other Notes

One unit equals the amount of enzyme required to turnover 1 μmole MUC per minute at pH 5.0 and 50 °C


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library