Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

F8180

Sigma-Aldrich

FMS (539-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

C-FMS, CD115, CSF1R, CSFR, FIM2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

18-24 nmol/min·mg

mol wt

~76 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CSF1R(1436)

General description

FMS, also called CSF1R colony stimulating factor 1 receptor, acts as a receptor for CSF1, and they both as key regulators of monocyte and macrophage functioning. It is a member of the platelet-derived growth factor (PDGF) family of proteins, and is a type III RTK (receptor tyrosine kinase). It is composed of five immunoglobulin (Ig)-like domains, a transmembrane region, a juxtamembrane domain, and kinase insert domain (KID) intersecting the kinase domain.

Biochem/physiol Actions

FMS also called CSF1R colony stimulating factor 1 receptor, and its ligand CSF1 are linked to poor prognosis in patients with solid tumors, and in patients with female reproductive cancers. In breast cancers with decreased claudin levels, this protein regulates the switch between proliferative and invasive state of the tumor, acting downstream of TGFβ (tumor growth factor β). Haploinsufficiency in this receptor might lead to aberration of microglial function, which might be a contributor to the pathogenesis of hereditary diffuse leukoencephalopathy with spheroids (HDLS).

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Sherr
International journal of cell cloning, 8 Suppl 1, 46-60 (1990-01-01)
Colony-stimulating factor-1 (CSF-1 or M-CSF) regulates pleiotropic developmental and functional responses of macrophages and their committed bone marrow progenitors and supports the viability of cells of the mononuclear phagocyte lineage. Its actions are mediated through its binding to cell surface
George Alexander Follows et al.
Oncogene, 24(22), 3643-3651 (2005-04-05)
The macrophage colony-stimulating factor receptor is encoded by the c-FMS gene, and it has been suggested that altered regulation of c-FMS expression may contribute to leukaemic transformation. c-FMS is expressed in pluripotent haemopoietic precursor cells and is subsequently upregulated during
Takuya Konno et al.
Neurology, 82(2), 139-148 (2013-12-18)
To clarify the genetic, clinicopathologic, and neuroimaging characteristics of patients with hereditary diffuse leukoencephalopathy with spheroids (HDLS) with the colony stimulating factor 1 receptor (CSF-1R) mutation. We performed molecular genetic analysis of CSF-1R in patients with HDLS. Detailed clinical and
Hang Liu et al.
Breast cancer research and treatment, 166(1), 95-107 (2017-07-22)
Endocrine resistance limits the efficacy of anti-estrogen therapies. Notch signaling is involved in modulating tumor-associated macrophage (TAM) differentiation and is upregulated in endocrine-resistant breast cancer cells. Here, we analyzed the role of Jagged1 in the regulation of TAM polarization to
Clare Pridans et al.
Scientific reports, 3, 3013-3013 (2013-10-23)
Hereditary diffuse leukoencephalopathy with spheroids (HDLS) in humans is a rare autosomal dominant disease characterized by giant neuroaxonal swellings (spheroids) within the CNS white matter. Symptoms are variable and can include personality and behavioural changes. Patients with this disease have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service