Skip to Content
MilliporeSigma

CB0500

CelLytic B Plus Kit

For bacterial lysis

Synonym(s):

Lysis and protein extraction kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106511
EC Number:
200-664-3
NACRES:
NA.56
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

usage

 kit sufficient for 50 g fresh or frozen cell paste lysis.

technique(s)

protein extraction: suitable

shipped in

wet ice

storage temp.

−20°C

General description

The CelLytic B Plus Kit is designed to efficiently lyse cells and extract proteins from both Gram negative, and difficult to lyse, Gram positive bacteria. This is accomplished using the standard CelLytic B, a proprietary non-ionic detergent in concert with lysozyme, Benzonase®, and protease inhibitors. This complete kit takes the guesswork out of protein extraction from a variety of bacterial species.

Application

This kit is compatible with FLAG®, HIS-Select®, and glutathione S-transferase based affinity chroma­tography protein purification systems. The detergent included in the CelLytic B Lysis Reagent can be removed from the protein by dialysis or ammonium sulfate precipitation, if necessary. The final purity of the recombinant product is usually higher than that obtained from traditional extraction methods. This is because the crude protein solution containing the non-ionic detergent eliminates much of the non-specific binding that occurs during chromatography. The mild detergent does not interfere with many enzyme assays or protein assays.

Features and Benefits

  • Lyse Gram positive and Gram negative bacteria
  • More efficient than sonication
  • Compatible with affinity purification
  • Isolate inclusion bodies for subsequent solubilization
  • Non-denaturing cell lysis preserves protein function

Other Notes

The CelLytic B Plus Kit contains all of the reagents and chemicals necessary to lyse both Gram-negative and Gram-positive bacteria. This kit also includes protease inhibitors to help prevent the proteolytic breakdown of proteins.

Legal Information

Benzonase® Nuclease is manufactured by Merck KGgA, Darmstadt, Germany and its affiliates. Nycomed Pharma A/S (Denmark) claims worldwide patent rights to Benzonase Nuclease, which are licensed exclusively to Merck KGaA, Darmstadt, Germany.
Covered by US Patent No 7,282,475 B2 and are sold for research use only. Commercial use requires addtional licenses.
Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
CelLytic is a trademark of Sigma-Aldrich Co. LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
HIS-Select is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under CelLytic B Plus Kit


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • CelLytic B Cell Lysis Reagent, For bacterial cell lysis, standard strength
    SDS

  • Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
    SDS

  • Protease Inhibitor Cocktail, for use in purification of Histidine-tagged proteins, DMSO solution
    SDS

  • Lysozyme from chicken egg white, 10 mg/mL
    SDS

pictograms

Health hazard

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library