Skip to Content
MilliporeSigma
All Photos(1)

Documents

909823

Sigma-Aldrich

3-Azido-D-alanine hydrochloride

≥95%

Synonym(s):

(R)-2-Amino-3-azidopropanoic acid hydrochloride, 3-Azido-D-alanine HCl, 3-Azido-D-alanine hydrochloride, H-D-Ala(N3)-OH*HCl, H-D-Aza-OH*HCl, H-D-Dap(N3)-OH*HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N4O2 · HCl
CAS Number:
Molecular Weight:
166.57
UNSPSC Code:
12352101

Assay

≥95%

form

powder

contains

≤11.0% water

reaction suitability

reaction type: click chemistry
reaction type: solution phase peptide synthesis

availability

available only in USA

storage temp.

2-8°C

Application

Some clickable unnatural D-amino acids can be fed to bacterial cells and incorporate into cell walls. The D-amino acids can also be used to generate L-RNA aptamers for the corresponding L-amino acid.

related product

Product No.
Description
Pricing

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service