Skip to Content
MilliporeSigma

Skip To

SML1639

BRD3308

≥98% (HPLC)

Synonym(s):

4-Acetamido-N-(2-amino-4-fluorophenyl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H14FN3O2
CAS Number:
Molecular Weight:
287.29
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(NC1=CC=C(C(NC2=CC=C(F)C=C2N)=O)C=C1)=O

InChI

1S/C15H14FN3O2/c1-9(20)18-12-5-2-10(3-6-12)15(21)19-14-7-4-11(16)8-13(14)17/h2-8H,17H2,1H3,(H,18,20)(H,19,21)

InChI key

RRJDFENBXIEAPD-UHFFFAOYSA-N

Biochem/physiol Actions

BRD3308 is a highly selective inhibitor of histone deacetylase 3 (HDAC3) with an IC50 value of 65 nM for HDAC3 vs. IC50 values of 1.08 μM and 1.15 μM for HDAC1 and HDAC2, respectively. BRD3308 protected pancreatic β cells, suppressing inflammatory cytokine-induced apoptosis and increasing insulin release without the toxicity associated with HDAC1 and HDAC2 inhibitors. In a rat model of type 2 diabetes, BRD3308 reduced hyperglycemia and increased insulin secretion without affecting weight gain. In another study, BRD3308 was found to activate HIV-1 transcription, disrupting HIV-1 latency.
BRD3308 is a highly selective inhibitor of histone deacetylase 3 (HDAC3).
BRD3308 promotes outgrowth of HIV-1 (human immunodeficiency virus 1) from inactive infected patient cells. It helps to increase β-cell proliferation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2682SML2736SML0268
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: >15 mg/mL

color

white to beige

color

faint brown to very dark brown-red

color

white to beige

color

white to beige


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters