Key Documents
EP0030124820
Eppendorf® PCR tube strips, 0.1 mL
with cap strips, flat, 0.1 mL, PCR clean, capacity 0.1 mL, with Cap Strips, flat (10x12 strips), pkg of 1000 ea
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
feature
PCR clean
with Cap Strips, flat (10x12 strips)
packaging
pkg of 1000 ea
manufacturer/tradename
Eppendorf® 0030124820
capacity
0.1 mL
0.1 mL
Features and Benefits
  • 8 reaction tubes in strip-format – ideal for small sample volumes
  • Easily adaptable for automation
  • Sealable using flat or domed strip-lids
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)