Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EMU041221

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Gpr109a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTTGCTTCTTGTGGGGTCTCACCATCGGCCTGACTGTCCACCTCCTCTATACAAACATGATGACCAAAAATGGCGAGGCATATCTGTGTAGCAGCTTCAGCATCTGTTACAACTTCAGGTGGCACGATGCTATGTTCCTCTTGGAATTCTTCTTGCCCCTGGCCATCATCTTGTTCTGCTCAGGCAGGATCATCTGGAGCCTGAGGCAGAGACAGATGGACAGACATGCCAAGATCAAGAGGGCCATCAACTTCATCATGGTGGTGGCTATTGTATTCATCATTTGCTTCCTACCCAGTGTGGCTGTGCGCATCCGCATCTTCTGGCTTCTCTACAAATATAACGTACGCAACTGTGACATCTACTCCTCGGTGGACCTGGCTTTCTTTACCACCCTTAGCTTTACCTACATGAACAGCATGCTGGACCCTGTGGTCTACTATTTCTCCAGCCCATCTTTCCCCAACTTCTTCTCCACGTGTATCAACCGCTGCCTTCGAAAGAAAACATTGGGTGAACCCGATA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Banabihari Giri et al.
International journal of molecular sciences, 20(18) (2019-09-22)
In this study, we used macrophage RAW264.7 cells to elucidate the molecular mechanism underlying the anti-inflammatory actions of niacin. Anti-inflammatory actions of niacin and a possible role of its receptor GPR109A have been studied previously. However, the precise molecular mechanism
Hideo Ohira et al.
Lipids in health and disease, 15(1), 213-213 (2016-12-13)
Interactions between adipocytes and macrophages are associated with metabolic disorders. Production of pro-inflammatory mediators and the release of free fatty acids (FFAs) increase when these cells are co-cultured; butyrate significantly diminishes these effects by suppressing both the macrophage inflammatory and
Genki Hayashi et al.
Human molecular genetics, 26(15), 2864-2873 (2017-05-02)
The induction of mitochondrial biogenesis could potentially alleviate mitochondrial and muscle disease. We show here that dimethyl fumarate (DMF) dose-dependently induces mitochondrial biogenesis and function dosed to cells in vitro, and also dosed in vivo to mice and humans. The
Samar Rezq et al.
The Journal of pharmacology and experimental therapeutics, 356(2), 456-465 (2015-12-02)
G protein-coupled receptor 109A (GPR109A) activation by its ligand nicotinic acid (NA) in immune cells increases Ca(2+) levels, and Ca(2+) induces glutamate release and oxidative stress in central blood pressure (BP)-regulating nuclei, for example, the rostral ventrolateral medulla (RVLM), leading

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service