Skip to Content
MilliporeSigma

8.55098

(3-Formylindolyl)acetamidomethyl polystyrene

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023
Form:
beads
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

aldehyde

storage temp.

2-8°C

General description

Versatile indole-based linker for the synthesis of secondary amides, carbamates, ureas [1,2,3,4] and aminopurines [5]. In a study comparing the cleavage rates of secondary amides, ureas, carbamates and sulfonamides from indole, dialkoxybenzaldehyde and Rink amide resins, the indole resin was found to be the most acid-labile [6].Reductive amination with excess of amine and NaBH(OAc)3 in DCE/TMOF [seepage], NaBH3CN in THF/TMOF/AcOH, or NaBH4-Ti(OiPr)4 in THF [2], converts the formyl group to the appropriate secondary amine. Cleavage can be effected with 2-5% TFA in DCM, depending on the nature of the product being released from the resin.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] K. G. Estep, et al. (1998) J. Org. Chem., 63, 5300.
[2] S. Bhattacharyya, et al. (2003) Tetrahedron Lett., 44, 6099.
[3] N. Hone, et al. (2003) Tetrahedron Lett., 44, 8169.
[4] L. Sereni, et al. (2004) Tetrahedron, 60, 8561.
[5] P. H. Dorff and R. S. Garigipati (2001) Tetrahedron Lett., 42, 2771.
[6] B. Yan, et al. (2000) J. Comb. Chem., 2, 66.

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined by elemental analysis of sulfur after coupling with tosylhydrazide): 0.60 - 1.20 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly (styrene-1% DVB), 100 - 200 mesh

Other Notes

Replaces: 01-64-0361

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
855098000504027269113704
855098002504027269113711
855098000104027269113698