Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

E6154

Sigma-Aldrich

Anti-phospho-Epidermal Growth Factor Receptor (pTyr1068) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-EGFR (pTyr1068)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen 170 kDa

species reactivity

human, rat

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EGFR(1956)

Related Categories

General description

The EGFR family includes four receptor tyrosine kinases, EGF R (ErbB1), ErbB2 (neu), ErbB3, and ErbB4. EGF R is present on many cell types of epithelial and mesenchymal lineages.
The gene EGFR (Epidermal Growth Factor Receptor) is mapped to human chromosome 7p11.2.

Immunogen

synthetic phosphopeptide derived from the region of EGFR that contains tyrosine 1068.

Application

Anti-phospho-EGFR [pTyr1068] antibody may be used at a working dilution of 1:1000 for immunoblotting using NIH3T3 cells expressing human EGF R, stimulated with EGF. The antibody has been in detection of EGF R [pTyr1068] in human lymphocytes.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The members of epidermal growth factor receptor (EGF R) or the ErbB receptor family have been identified as useful biomarkers and targets for cancer therapy. EGF R binds EGF and induces tyrosine phosphorylation leading to proliferation of cells. EGF R is capable of binding transforming growth factor-a and heparin-binding EGF in addition to EGF. There are numerous effector molecule activated by EGF R that result in a variety of biological processes such as morphogenesis, cell motility, apoptosis, differentiation and organ repair and maintenance. Deregulation of EGF R signaling is implicated in progression of a wide variety of tumors, invasion and metastasis. Phosphorylation of Tyr1068 occurs by autophosphorylation of EGF R and is required for the binding of Grb2 and activation of ERK1/2 pathway Anti-phospho-EGFR [pTyr1068] specifically recognizes human and rat EGFR phosphorylated at tyrosine 1068.

Physical form

Solution in Dulbecco′s phosphate buffered saline, with 50% glycerol, 1 mg/ml BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian P Rybak et al.
PloS one, 8(4), e61716-e61716 (2013-04-27)
Prostate cancer stem-like cells (PCSCs) are being intensely investigated largely owing to their contributions towards prostate tumorigenesis, however, our understanding of PCSC biology, including their critical pathways, remains incompletely understood. While epidermal growth factor (EGF) is widely used in maintaining
Gergely Keszler et al.
Archives of biochemistry and biophysics, 436(1), 69-77 (2005-03-09)
The inborn deficiency of adenosine deaminase is characterised by accumulation of excess amounts of cytotoxic deoxyadenine nucleotides in lymphocytes. Formation of dATP requires phosphorylation of deoxyadenosine by deoxycytidine kinase (dCK), the main nucleoside salvage enzyme in lymphoid cells. Activation of
Targeting the EGFR signaling pathway in cancer therapy
Seshacharyulu P et al
Expert Opinion on Therapeutic Targets, 16, 15-31 (2010)
L Lanzetti et al.
Nature, 408(6810), 374-377 (2000-12-01)
How epidermal growth factor receptor (EGFR) signalling is linked to EGFR trafficking is largely unknown. Signalling and trafficking involve small GTPases of the Rho and Rab families, respectively. But it remains unknown whether the signalling relying on these two classes
Chromosome 7p11.2 (EGFR) variation influences glioma risk.
Sanson M, et al.
Human Molecular Genetics, 20(14), 2897-2904 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service