E3511
cis-8,11,14-Eicosatrienoic acid methyl ester
≥99%
Synonym(s):
Dihomo-γ-linolenic acid methyl ester, Fame 20:3n-6, Methyl DGLA
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C21H36O2
CAS Number:
Molecular Weight:
320.51
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
biological source
synthetic
Assay
≥99%
form
liquid
functional group
ester
lipid type
omega FAs
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCC\C=C/C\C=C/C\C=C/CCCCCCC(=O)OC
InChI
1S/C21H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h7-8,10-11,13-14H,3-6,9,12,15-20H2,1-2H3/b8-7-,11-10-,14-13-
InChI key
QHATYOWJCAQINT-JPFHKJGASA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Sealed ampule
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service