Skip to Content
MilliporeSigma

Skip To

E6001

cis-11,14,17-Eicosatrienoic acid methyl ester

≥98%, liquid

Synonym(s):

Methyl 11,14,17-eicosatrienoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H36O2
CAS Number:
Molecular Weight:
320.51
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥98%

form

liquid

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CC\C=C\C\C=C\C\C=C\CCCCCCCCCC(=O)OC

InChI

1S/C21H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h4-5,7-8,10-11H,3,6,9,12-20H2,1-2H3/b5-4+,8-7+,11-10+

InChI key

XQAVRBUXEPJVRC-JSIPCRQOSA-N

Packaging

Sealed ampule.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E3511D2659D4034
description

≥98%, liquid

description

≥99%

description

≥98%

description

≥98%

biological source

synthetic (organic)

biological source

synthetic

biological source

synthetic (organic)

biological source

synthetic (organic)

functional group

ester

functional group

ester

functional group

ester

functional group

ester

assay

≥98%

assay

≥99%

assay

≥98%

assay

≥98%

form

liquid

form

liquid

form

liquid

form

liquid

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

wgk

WGK 3

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters