Skip to Content
MilliporeSigma
All Photos(1)

Documents

E7877

Sigma-Aldrich

cis-11,14-Eicosadienoic acid methyl ester

≥98% (GC)

Synonym(s):

Methyl cis,cis-11,14-eicosadienoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H38O2
CAS Number:
Molecular Weight:
322.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

plant oil (Safflower)

Assay

≥98% (GC)

form

liquid

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C\C\C=C\CCCCCCCCCC(=O)OC

InChI

1S/C21H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h7-8,10-11H,3-6,9,12-20H2,1-2H3/b8-7+,11-10+

InChI key

GWJCFAOQCNNFAM-ZDVGBALWSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sealed ampule.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service