Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS7247

Corning® non-treated 1536 well microplates

white, high base, w/o lid, non-sterile

Synonym(s):

1536 multiwell plates, 1536 well microplates, 1536 well microtiter plates, 1536 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
polystyrene plate
square wells
white
white bottom
size:
1536 wells
well volume:
12.5 μL
well working volume:
1-8 μL

material

polystyrene plate
square wells
white
white bottom

sterility

non-sterile

feature

lid: no
skirt

packaging

case of 50 ea

manufacturer/tradename

Corning 7247

size

1536 wells

well volume

12.5 μL

well working volume

1-8 μL

color

white

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service