Skip to Content
MilliporeSigma

M8562

Greiner CELLSTAR® multiwell culture plates

6 wells (TC treated with lid)

Synonym(s):

culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates, Greiner 657160

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat clear wells, polystyrene
Size:
6 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat clear wells, polystyrene

sterility

sterile

feature

lid

packaging

pack of 1 ea

manufacturer/tradename

Greiner 657160

size

6 wells

well volume

16 mL

well working volume

2- 5 mL

binding type

Tissue Culture (TC)-treated surface

General description

The Greiner CELLSTAR® multiwell culture plate is a 6-well clear, polystyrene plate designed for cell culture. the surface has been Tissue Culture (TC)- treated and includes a lid with condensation rings. Each plate is sterile and single-packed for easy use.

Application

Greiner CELLSTAR® multiwell culture plates have been used:
  • in neuromast damage protocol and the basic chemically induced inflammation assay (ChIn) using zebrafish larvae
  • to prepare karaya gum (KG)/poly(vinyl alcohol) (PVA) cryogel for the infiltration of stem cells
  • for the purification of dendritic cells (DC) from mice spleen

Features and Benefits

  • Tissue Culture (TC)-treated surface for enhanced cell adhesion
  • Free of detectable DNase, RNase, human DNA Non-Cytotoxic
  • Single position lids to prevent cross-contamination
  • High clarity and low autofluorescence for clear observation of cell cultures
  • Contains single position lids to prevent cross-contamination
  • Lid enables optimal gas exchange with minimal evaporation
  • Alphanumeric well coding for easy identification and tracking
  • Seamless integration with automated systems
  • Convenient, easy-to-open packaging
  • Growth area per well: 9.6 cm2
  • Working volume per well: 2 ml - 5 ml
  • Sterile and non-pyrogenic

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library