Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
material
polycarbonate , round bottle, screw top cap, translucent bottle, translucent polycarbonate , translucent polypropylene screw closure
joint
Joints threaded neck (35.8)
feature
RCF 13,700 × g, with cap
packaging
case of 24, pack of 4
manufacturer/tradename
Nalgene® 3122-0500
parameter
13700 × g max. RCF
bottle diam. × L
61.77 mm × 125.2 mm , 69.52 mm × 168.9 mm
nominal capacity
500 mL
General description
Features and Benefits
- Bottles are autoclavable.
- Have excellent mechanical strength properties.
- Easy visualization of samples.
- Ideal for low-speed centrifugation of biological and chemical samples.
- To ensure enhanced leakproof security during high-speed spins or when dealing with hazardous materials, pair the bottles with polypropylene sealing cap assemblies.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| feature RCF 13,700 × g, with cap | feature RCF 7,100 × g, with cap | feature RCF 27,500 × g, with cap (sealing) | feature RCF 13,700 × g, with cap |
| material polycarbonate , screw top cap, translucent polycarbonate , round bottle, translucent polypropylene screw closure, translucent bottle | material polycarbonate , round bottle, screw top cap, translucent bottle, translucent polycarbonate , translucent polypropylene screw closure | material polycarbonate , round bottle, screw top cap (Sealing), translucent bottle, translucent polycarbonate , translucent polypropylene screw closure | material polypropylene copolymer bottle, round bottle, screw top cap, translucent bottle |
| manufacturer/tradename Nalgene® 3122-0500 | manufacturer/tradename Nalgene® 3122-1000 | manufacturer/tradename Nalgene® 3122-0250 | manufacturer/tradename Nalgene® 3120-9500 |
| packaging case of 24, pack of 4 | packaging case of 16, pack of 4 | packaging pack of 4 | packaging pack of 4 |
| nominal capacity 500 mL | nominal capacity 1000 mL | nominal capacity 250 mL | nominal capacity 500 mL |
| bottle diam. × L 61.77 mm × 125.2 mm | bottle diam. × L 98.17 mm × 169.7 mm | bottle diam. × L 61.77 mm × 125.2 mm | bottle diam. × L 69.52 mm × 168.9 mm |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge bottles, Style 3122
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

