Key Documents
CLS431463
Corning® flask, canted neck, 25 cm, vented
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
UNSPSC Code:
41122104
NACRES:
NB.13
material
canted neck
high-density polyethylene cap
polystyrene
rectangular bottom flask
vent cap
sterility
sterile; γ-irradiated
feature
cap
closure type Vented cap
packaging
pack of 20
case of 200
manufacturer/tradename
Corning 431463
surface area
25 cm2
working volume
5-7.5 mL
binding type
non-treated surface
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)