Skip to Content
MilliporeSigma

M4811

Greiner high and medium binding 96 well plates

flat bottom white wells, medium binding surface, non-sterile

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well strip plates, Greiner 655075

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
flat bottom white wells, white polystyrene
Size:
96 wells
Sterility:
non-sterile
Binding type:
medium binding surface
Feature:
lid: no, skirt (F-bottom)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom white wells, white polystyrene

sterility

non-sterile

feature

lid: no, skirt (F-bottom)

packaging

case of 40

manufacturer/tradename

Greiner 655075

maximum volume

392 μL

size

96 wells

working volume

340 μL

binding type

medium binding surface

General description

LUMITRAC 200 96W Microplate, well working volume 25-340 μL; total well volume 392 μL.
Polystyrene Multiwell plates from Greiner for immunoassay and high throughput screening applications. High binding plates are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding plates is suggested.

Application

White plates minimize cross talk in luminescence procedures. LUMITRAC 200 will bind up to 200 ng/cm2. Free of detectable DNase/RNase, human DNA and pyrogens.

Other Notes

96 well flat (F) bottom. medium binding microplate, without lid, Chimney Style; alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Legal Information

LUMITRAC is a trademark of Greiner Bio-One GmbH


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library