Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS3917

Corning® 96 Well Solid Polystyrene Microplate

flat bottom, white polystyrene, Tissue Culture (TC)-treated surface, bag of 20, lid

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
flat bottom
white bottom
white polystyrene
size:
96 wells
Pricing and availability is not currently available.

material

flat bottom
white bottom
white polystyrene

sterility

sterile

feature

lid
skirt

packaging

bag of 20

manufacturer/tradename

Corning 3917

size

96 wells

well volume

360 μL

working volume

75-200 μL

color

white

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Solid Polystyrene Microplate have Flat bottom with a working volume of 75 to 200 μL.
  • Opaque white polystyrene walls.
  • Display low background luminescence and fluorescence.
  • Non-reversible lids with condensation ring to reduce contamination.
  • Nonpyrogenic and sterilized by gamma radiation.
  • Individual alphanumeric codes enable well identification.
  • Hydrophilic surface and negatively charged optimally for cell attachment and growth.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service