Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

CLS353263

Corning® Falcon® 96 Well Library Storage Plate

conical clear polypropylene, sterile

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122102
NACRES:
NB.13
material:
conical clear polypropylene
sterility:
sterile

material

conical clear polypropylene

sterility

sterile

manufacturer/tradename

Corning 353163

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for: Compound storage

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service