Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CLS3355

Corning® 96 Well Storage Microplate

round bottom, white polypropylene, bag of 25 ×, non-sterile, lid: no

Synonym(s):

96 multiwell plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

round bottom
white bottom
white polypropylene

sterility

non-sterile

feature

lid: no

manufacturer/tradename

Corning 3355

packaging

bag of 25 ×

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

white

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Corning® 96 well microplates support various assay and storage application requirements.
  • Features uniform skirt heights for the greater robotic gripping surface.
  • Resistant to most solvents including DMSO, ethanol, methanol.
  • RNase-/DNase-free.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service