Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

COR L303

96020722, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR L303, 96020722

biological source

human lung

growth mode

Aggregates in suspension

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human lung small cell carcinoma

Cell Line Description

COR L303 cells were isolated from a female patient of unknown age with small cell lung carcinoma. The sample was taken pre-therapy.

Culture Medium

RPM1 1640 + 10% Foetal Bovine Serum (FBS) + 2mM Glutamine.

Subculture Routine

Maintain cultures at 3-9 x 100,000/ml, 37°C, 5% CO2

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service