COR-L279
96020724, human lung (lymph node metastasis), Not specified
Synonym(s):
CORL279 Cells, LuCL4 Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Product Name
COR-L279, 96020724
biological source
human lung (lymph node metastasis)
growth mode
Aggregates in suspension
karyotype
Not specified
morphology
Not specified
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
metastasis
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human lung small cell carcinoma
Cell Line Description
COR-L279 has been isolated from a lymph node of an untreated 63 year-old male. The degenerated and necrotic morphology of the sample obtained was consistent with metastatic small cell carcinoma of the lung.
Application
Cytotoxicity studies, study of small cell lung cancer
DNA Profile
STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11
D13S317: 12
D16S539: 11
D5S818: 12
D7S820: 9,10
THO1: 6,9.3
TPOX: 9,11
vWA: 16,17
CSF1PO: 11
D13S317: 12
D16S539: 11
D5S818: 12
D7S820: 9,10
THO1: 6,9.3
TPOX: 9,11
vWA: 16,17
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service