Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M15

95102517, mouse kidney, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

M15, 95102517

biological source

mouse kidney

growth mode

Adherent

karyotype

Hyperdiploid

morphology

Epithelial

products

WT1 (expressed gene)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse mesonephric epithelium, polyoma virus large T transformed

Cell Line Description

The WT1-expressing mesonephric cell line M15 (alias Meso15) was established from mouse mesonephros transgenically expressing the large T protein of polyoma virus under the control of the early viral enhancer. As a tumour suppresser gene with a key role in urogenital development, WT1 is implicated as predisposition gene in the pathogenesis of Wilms′ tumour (WT).

Application

Gene expression and protein studies connected to kidney development and Wilms′ tumourigenesis.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:5 to 1:10 i.e. seeding at 5x1,000 to 1x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. NB: Cell growth is impaired at lower densities.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service