Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RAG

89040605, mouse kidney, Ameboid-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RAG, 89040605

biological source

mouse kidney

growth mode

Adherent

karyotype

Modal no. 68, hypertriploid

morphology

Ameboid-like

products

ES-2

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c renal adenocarcinoma

Cell Line Description

Derived from the original Renal-2a BALB/c renal adenocarcinoma. Cells produce the unique kidney specific esterase ES-2 and have been used in somatic cell hybridisation studies because of their azaguanine resistant properties.

Application

Hybridisation, resistance and tumourigenicity studies

Culture Medium

EMEM (HBSS) + 2mM Glutamine + 2% Non Essential Amino Acids (NEAA) + 2% Vitamins + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:3 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service