Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M-1

95092201, mouse kidney, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

M-1, 95092201

biological source

mouse kidney

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse kidney cortical collecting duct, SV40 transformed

Cell Line Description

Derived from the cortical collecting duct (CCD) of a mouse, transgenic for the early region of SV40 (Tg(SV40E)Bri/7). They express many characteristics of the CCD like epithelial morphology and CCD-specific antigens. M-1 cells grown on permeable supports exhibited a high transepithelial resistance concomitant with the development of a lumen-negative transepithelial potential difference. The M-1 cell line and sub-clones exhibit principal cell (PC) functions and ß-intercalated cell (ß-ICC) functions, both typical for the heterogeneous epithelium found in the renal collecting duct. M-1 cells and sub-clones show a heterogeneous expression of PC and ß-ICC antigens with 5-10% exhibiting a dual PC/ß-ICC phenotype.

Application

Kidney cell studies, differentiation studies.

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 5μm Dexamethasone (DXMT) + 5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Prolonged incubation with trypsin may be necessary for subculture.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service