Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CCO

95060212, fish ovary, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CCO, 95060212

biological source

fish ovary

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Channel Catfish ovary

Cell Line Description

CCO was established from the ovaries of a healthy, juvenile channel catfish, Ictalurus punctatus. The cells grow at temperatures from 10°C to 35°C with the optimum at 30°C and with a reported population doubling up to 12 hours. CCO cells have been shown to replicated channel catfish virus but were refractory to 12 other viruses from fish, birds and mammals.

Application

Isolation of channel catfish virus

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 30°C. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 and culture for at least one week with 1-2 media ch

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service