Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CAR

89072611, fish fin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

CAR, 89072611

biological source

fish fin

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Goldfish fin tissue

Cell Line Description

The cells support the growth of grunt fin agent with CPE at 20°C and 25°C. Also susceptible to infectious pancreatic necrosis virus of trout at 28°C.

Application

Growth factors and virus studies: Infectious pancreatic necrotic virus

Culture Medium

BME (HBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 using 0.25% trypsin; 5% CO2; 25°C. Fish cell lines detach easily during transit if the culture is too young. After resuscitation split cells 1:3 to 1:4 and culture for at least one week with 1-2 media ch

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service