Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RTG-2

90102529, fish ovary, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RTG-2, 90102529

biological source

fish ovary

growth mode

Adherent

karyotype

2n = 60

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Trout Rainbow gonad tissue

Cell Line Description

From pooled male/female gonad tissue of yearling rainbow trout (Oncorhynchus mykiss). First cell line to be established from cold-blooded vertebrates. 8-fold difference in growth rate in temperature range 4-24°C. Susceptible to infectious pancreatic necrosis virus, Egtved virus and arboviruses (EEE, VEE).

Application

Virus studies: Inf. pacr. necrotic virus, Egtved virus, arboviruses (EEE, VEE)

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Passage using 0.25% trypsin below 24°C. Cells may be seeded as few as 3x10,000 cells/ml 5% CO2; 24°C. Fish cell lines detach easily during transit if the culture is very youn

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service