Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AT6.1

94101452, rat prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

AT6.1, 94101452

biological source

rat prostate

growth mode

Adherent

karyotype

44XY, +4, + 12 del(3)(3q32-36)del(15)p14

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat dorsal prostatic adenocarcinoma

Cell Line Description

A highly metastatic, anaplastic, androgen-independent sub-line which spontaneously developed during the serial passage of the non-metastatic, well-differentiated, androgen-responsive R-3327-H sub-lines in the laboratory of Dr J Schalken, Nijmegan, The Netherlands.

Application

Used in studies in which micro-cell mediated chromosomal transfer of human chromosome 11 suppressed metastatic ability. This suppresser function was localised to 11p11.2-13.

Culture Medium

RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service