Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

K1 Cell Line human

92030501, human thyroid, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

K1 Cell Line human, 92030501

biological source

human thyroid

description

Human thyroid carcinoma

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Wild-type p53 tumour suppressor gene

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human thyroid carcinoma

Cell Line Description

Derived from a primary papillary thyroid carcinoma. Retains thyroid follicular cell differentiation e.g. thyroglobulin synthesis. Expresses wild-type p53 tumour suppresser gene. It has been reported that K1 cells have their origin in the thyroid papillary carcinoma cell line GLAG-66.

Application

K1 Cell Line human has also been used to study and correlate the expression levels of c-KIT (a receptor tyrosine kinase) and two markers of thyrocyte differentiation (PAX8 and TTF-1).
Tumourigenicity studies, particularly the role of p53

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 11,12
D13S317: 11,14
D16S539: 11,12
D5S818: 10,11
D7S820: 11
THO1: 6,9
TPOX: 8
vWA: 17,18

Culture Medium

DMEM:Ham′s F12:MCDB 105 (2:1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2x1,000,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service