Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Hs 27

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human prostate

packaging

tube of 5 μg 94041901-DNA-5UG
pkg of vial of cells 94041901-1VL

growth mode

Adherent

karyotype

46XY

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human foreskin, fibroblast

Cell Line Description

Hs27 is one of a series of human foreskin fibroblast lines developed at NBL, California. The material was obtained from a normal new-born, and the cells possess the G6PD type A phenotype. Longevity studies carried out at NBL demonstrated that the cells can be propagated for 42 passages. There were no consistent aberrations in analysed cells. Ethnicity: Black.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service