Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

TGP 52

94022429, mouse pancreas, Trabecular

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

TGP 52, 94022429

biological source

mouse pancreas

growth mode

Adherent

karyotype

Not specified

morphology

Trabecular

products

Somatostatin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse islet cell tumour

Cell Line Description

TGP 52 has been isolated from an islet tumour arising in the pancreas of a transgenic Tg (E1a-1-SV40E) Bri18 mouse. These mice carry a pseudogene construct composed of a pancreatic specific elastase-1 promoter linked to the SV40T antigen and thereby represent an animal model for pancreatic carcinogenesis. TGP 52 cells are SV40T antigen positive and secrete somatostatin, it is one of a series of pancreatic tumour lines deposited at ECACC. Cells transiently produce insulin in culture followed by somatostatin.

Application

Study of endocrine pancreas carcinomas, hormone assays.

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service